Assay Design



Gene Transcripts

Gene Symbol
NOM1
Entrez Gene ID
64434
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
C7orf3, PPP1R113, SGD1
Chromosome Location
Chr.7: 156949723 - 156973182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138400.1 NP_612409.1 8 - 9 2182 77
GenBank mRNA AY576779.1 - 8 - 9 2182 77
BC007902.2 - 4 - 5 473 77

View Full Product Details