Product Details

Assay Reference Genome
Location

Chr.4:41263316 on build GRCh38
Cytoband
4p13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
7345
Gene Symbol
UCHL1
Gene Name
ubiquitin C-terminal hydrolase L1
Gene Aliases
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
Location
Chr.4:41256881-41268429 on build GRCh38
Assay Gene Location
Overlaps Exon 7 - Intron 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
UCHL1 NM_004181.4 NP_004172.2
AK054579.1
AK315368.1
BC000332.2 AAH00332.1
BC005117.1 AAH05117.1
BC006305.2 AAH06305.1
FJ224355.1
GQ472201.1
X04741.1 CAA28443.1

View Full Product Details