Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243224.1 | NP_001230153.1 | 1 - 2 | 308 | 55 |
NM_004584.2 | NP_004575.1 | 4 - 5 | 444 | 55 | |
XM_006718652.3 | XP_006718715.1 | 2 - 3 | 656 | 55 | |
XM_017018098.1 | XP_016873587.1 | 2 - 3 | 184 | 55 | |
GenBank mRNA | AK310213.1 | - | 1 - 2 | 731 | 55 |
AK315348.1 | - | 4 - 5 | 383 | 55 | |
BC014848.1 | - | 4 - 5 | 444 | 55 | |
CB164642.1 | - | 1 - 2 | 308 | 55 | |
CN402466.1 | - | 1 - 2 | 273 | 55 | |
CR536508.1 | - | 4 - 5 | 351 | 55 | |
U53174.1 | - | 4 - 5 | 427 | 55 |