Assay Design



Gene Transcripts

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
Chromosome Location
Chr.4: 113451032 - 113761927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001221.3 NP_001212.2 1 - 2 925 94
NM_001321566.1 NP_001308495.1 1 - 2 925 94
NM_001321567.1 NP_001308496.1 1 - 2 925 94
NM_001321568.1 NP_001308497.1 1 - 2 925 94
NM_001321569.1 NP_001308498.1 1 - 2 925 94
NM_001321570.1 NP_001308499.1 1 - 2 925 94
NM_001321571.1 NP_001308500.1 1 - 2 925 94
NM_001321572.1 NP_001308501.1 1 - 2 925 94
NM_001321573.1 NP_001308502.1 1 - 2 925 94
NM_001321574.1 NP_001308503.1 1 - 2 925 94
NM_001321575.1 NP_001308504.1 1 - 2 925 94
NM_001321576.1 NP_001308505.1 1 - 2 925 94
NM_001321577.1 NP_001308506.1 1 - 2 925 94
NM_001321578.1 NP_001308507.1 1 - 2 925 94
NM_001321579.1 NP_001308508.1 1 - 2 925 94
NM_001321580.1 NP_001308509.1 1 - 2 925 94
NM_001321581.1 NP_001308510.1 1 - 2 925 94
NM_001321582.1 NP_001308511.1 1 - 2 925 94
NM_001321583.1 NP_001308512.1 1 - 2 925 94
NM_001321584.1 NP_001308513.1 1 - 2 925 94
NM_001321585.1 NP_001308514.1 1 - 2 925 94
NM_001321586.1 NP_001308515.1 1 - 2 925 94
NM_001321587.1 NP_001308516.1 1 - 2 925 94
NM_001321588.1 NP_001308517.1 1 - 2 925 94
NM_001321589.1 NP_001308518.1 1 - 2 925 94
NM_001321590.1 NP_001308519.1 1 - 2 925 94
NM_001321591.1 NP_001308520.1 1 - 2 925 94
NM_001321592.1 NP_001308521.1 1 - 2 925 94
NM_172114.1 NP_742112.1 1 - 2 925 94
NM_172115.2 NP_742113.1 1 - 2 925 94
NM_172127.2 NP_742125.1 1 - 2 925 94
NM_172128.2 NP_742126.1 1 - 2 925 94
NM_172129.1 NP_742127.1 1 - 2 925 94
XM_005263253.3 XP_005263310.1 1 - 2 743 94
XM_005263254.3 XP_005263311.1 1 - 2 764 94
XM_006714330.2 XP_006714393.1 1 - 2 764 94
XM_011532289.1 XP_011530591.1 1 - 2 743 94
XM_011532290.1 XP_011530592.1 1 - 2 743 94
XM_011532291.1 XP_011530593.1 1 - 2 743 94
XM_011532295.1 XP_011530597.1 1 - 2 764 94
XM_017008672.1 XP_016864161.1 1 - 2 764 94
XM_017008673.1 XP_016864162.1 1 - 2 764 94
XM_017008674.1 XP_016864163.1 1 - 2 764 94
XM_017008675.1 XP_016864164.1 1 - 2 764 94
XM_017008677.1 XP_016864166.1 1 - 2 764 94
XM_017008678.1 XP_016864167.1 1 - 2 743 94
GenBank mRNA AB209288.1 - 1 - 2 561 94
AJ252239.1 - 1 - 2 499 94
AK055642.1 - 1 - 2 443 94
AK294517.1 - 1 - 2 216 94
AY987011.1 - 1 - 2 66 94
BC022558.1 - 1 - 2 407 94
BC032784.1 - 1 - 2 128 94

View Full Product Details