Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024807.2 | NP_001019978.1 | 14 - 15 | 1847 | 70 |
NM_005166.4 | NP_005157.1 | 14 - 15 | 1844 | 70 | |
XM_017026737.1 | XP_016882226.1 | 13 - 14 | 1688 | 70 | |
XM_017026738.1 | XP_016882227.1 | 13 - 14 | 1685 | 70 | |
GenBank mRNA | AK092376.1 | - | 14 - 15 | 1647 | 70 |
AK297339.1 | - | 13 - 14 | 1657 | 70 | |
AK297640.1 | - | 14 - 15 | 1543 | 70 | |
AK312292.1 | - | 14 - 15 | 1714 | 70 | |
BC012889.1 | - | 14 - 15 | 1721 | 70 | |
BC013850.1 | - | 14 - 15 | 1730 | 70 | |
EF036503.1 | - | 3 - 4 | 220 | 70 | |
U48437.1 | - | 14 - 15 | 1693 | 70 |