Assay Design



Gene Transcripts

Gene Symbol
COL3A1
Entrez Gene ID
1281
Gene Name
collagen type III alpha 1 chain
Gene Aliases
EDS4A
Chromosome Location
Chr.2: 188974373 - 189012746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000090.3 NP_000081.1 14 - 15 1113 112
GenBank mRNA AK308010.1 - 10 - 11 878 112
BC028178.1 - 14 - 15 1111 112
S79877.1 - 2 - 3 94 112
X14420.1 - 14 - 15 1098 112
X15332.1 - 10 - 11 555 112

View Full Product Details