Assay Design



Gene Transcripts

Gene Symbol
COL3A1
Entrez Gene ID
1281
Gene Name
collagen type III alpha 1 chain
Gene Aliases
EDS4A
Chromosome Location
Chr.2: 188974373 - 189012746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000090.3 NP_000081.1 16 - 17 1268 61
GenBank mRNA BC028178.1 - 16 - 17 1266 61
X14420.1 - 16 - 17 1253 61
X15332.1 - 12 - 13 710 61

View Full Product Details