Assay Design



Gene Transcripts

Gene Symbol
COL3A1
Entrez Gene ID
1281
Gene Name
collagen type III alpha 1 chain
Gene Aliases
EDS4A
Chromosome Location
Chr.2: 188974373 - 189012746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000090.3 NP_000081.1 46 - 47 3533 94
GenBank mRNA X01655.1 - 3 - 4 224 94
X06700.1 - 7 - 8 569 94
X14420.1 - 46 - 47 3518 94
X15332.1 - 42 - 43 2975 94

View Full Product Details