Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000090.3 | NP_000081.1 | 48 - 49 | 3940 | 65 |
GenBank mRNA | AK091853.1 | - | 1 - 2 | 920 | 65 |
BC028178.1 | - | 38 - 39 | 3029 | 65 | |
X01742.1 | - | 1 - 2 | 235 | 65 | |
X06700.1 | - | 9 - 10 | 976 | 65 | |
X14420.1 | - | 48 - 49 | 3925 | 65 |