Assay Design



Gene Transcripts

Gene Symbol
OR5AP2
Entrez Gene ID
338675
Gene Name
olfactory receptor family 5 subfamily AP member 2
Gene Aliases
OR9J1
Chromosome Location
Chr.11: 56641489 - 56642439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002925.1 NP_001002925.1 1 - 1 625 93
GenBank mRNA BC136989.1 - 1 - 1 657 93
BC136990.1 - 1 - 1 657 93

View Full Product Details