Assay Design



Gene Transcripts

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
Chromosome Location
Chr.3: 97821930 - 97944986 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153605.3 NP_705833.3 15 - 16 8167 66
XM_005247117.4 XP_005247174.1 14 - 15 7870 66
GenBank mRNA AF451988.1 - 3 - 4 296 66
AK091199.1 - 7 - 8 638 66
AL831927.1 - 3 - 4 190 66
BX647079.1 - 7 - 8 655 66
BX648353.1 - 12 - 13 2217 66
CR749313.1 - 12 - 13 1292 66

View Full Product Details