Assay Design



Gene Transcripts

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
Chromosome Location
Chr.3: 97821930 - 97944986 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153605.3 NP_705833.3 18 - 19 8547 119
XM_005247117.4 XP_005247174.1 17 - 18 8250 119
GenBank mRNA AF451988.1 - 6 - 7 664 119
AK091199.1 - 10 - 11 1018 119
AK098569.1 - 2 - 3 339 119
AL831927.1 - 6 - 7 570 119
BC036247.1 - 1 - 2 90 119
BC050394.1 - 3 - 4 330 119
BX647079.1 - 10 - 11 1035 119
BX648353.1 - 15 - 16 2597 119
CR749313.1 - 15 - 16 1672 119

View Full Product Details