Assay Design



Gene Transcripts

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
Chromosome Location
Chr.3: 97821930 - 97944986 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153605.3 NP_705833.3 19 - 20 8673 65
XM_005247117.4 XP_005247174.1 18 - 19 8376 65
GenBank mRNA AF451988.1 - 7 - 8 790 65
AK091199.1 - 11 - 12 1144 65
AK098569.1 - 3 - 4 465 65
AL831927.1 - 7 - 8 696 65
BC036247.1 - 2 - 3 216 65
BX647079.1 - 11 - 12 1161 65
BX648353.1 - 16 - 17 2723 65
CR749313.1 - 16 - 17 1798 65

View Full Product Details