Assay Design



Gene Transcripts

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
Chromosome Location
Chr.3: 97821930 - 97944986 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153605.3 NP_705833.3 20 - 21 8828 65
XM_005247117.4 XP_005247174.1 19 - 20 8531 65
GenBank mRNA AF451988.1 - 8 - 9 945 65
AK091199.1 - 12 - 13 1299 65
AK098569.1 - 4 - 5 620 65
AL831927.1 - 8 - 9 851 65
BC050394.1 - 5 - 6 611 65
BX647079.1 - 12 - 13 1316 65
BX648353.1 - 17 - 18 2878 65
CR749313.1 - 17 - 18 1953 65

View Full Product Details