Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001553.2 | NP_001544.1 | 4 - 5 | 128 | |
GenBank mRNA | AI761388.1 | - | 1 - 2 | 272 | 129 |
BC017201.2 | - | 4 - 5 | 128 | ||
BC066339.1 | - | 4 - 5 | 128 | ||
S75725.1 | - | 4 - 5 | 128 |