Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015949.2 | NP_057033.2 | 4 - 5 | 554 | 98 |
GenBank mRNA | AF132954.1 | - | 4 - 5 | 441 | 98 |
AK023560.1 | - | 4 - 5 | 478 | 98 | |
AK024305.1 | - | 3 - 4 | 1255 | 98 | |
AK097899.1 | - | 3 - 4 | 2215 | 98 | |
AK125863.1 | - | 3 - 4 | 2820 | 98 | |
AL133014.1 | - | 3 - 4 | 1344 | 98 | |
BC003550.1 | - | 4 - 5 | 496 | 98 |