Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015949.2 | NP_057033.2 | 5 - 6 | 698 | 74 |
GenBank mRNA | AF132954.1 | - | 5 - 6 | 585 | 74 |
AK023560.1 | - | 5 - 6 | 622 | 74 | |
AK024305.1 | - | 4 - 5 | 1399 | 74 | |
AK097899.1 | - | 4 - 5 | 2359 | 74 | |
AK125863.1 | - | 4 - 5 | 2964 | 74 | |
AL133014.1 | - | 4 - 5 | 1488 | 74 | |
BC003550.1 | - | 5 - 6 | 640 | 74 |