Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079675.2 | NP_001073143.1 | 2 - 3 | 361 | 53 |
NM_001986.2 | NP_001977.1 | 2 - 3 | 190 | 53 | |
XM_017024349.1 | XP_016879838.1 | 2 - 3 | 152 | 53 | |
GenBank mRNA | AK290429.1 | - | 2 - 3 | 152 | 53 |
AK298872.1 | - | 1 - 2 | 158 | 53 | |
AK312934.1 | - | 2 - 3 | 152 | 53 | |
BC016623.1 | - | 2 - 3 | 132 | 53 | |
BM013535.1 | - | 2 - 3 | 190 | 53 | |
BP268896.1 | - | 2 - 3 | 268 | 53 | |
DC375462.1 | - | 2 - 3 | 362 | 53 | |
DR156106.1 | - | 2 - 3 | 358 | 53 | |
EU693079.1 | - | - | 262 | 53 | |
U18018.1 | - | 2 - 3 | 265 | 53 |