Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016329.3 | NP_057413.2 | 13 - 14 | 1810 | 58 |
XM_005265221.3 | XP_005265278.1 | 13 - 14 | 1926 | 58 | |
XM_006713203.3 | XP_006713266.1 | 14 - 15 | 2048 | 58 | |
XM_006713204.3 | XP_006713267.1 | 13 - 14 | 1748 | 58 | |
XM_011533824.2 | XP_011532126.1 | 14 - 15 | 1924 | 58 | |
XM_011533825.2 | XP_011532127.1 | 14 - 15 | 1771 | 58 | |
GenBank mRNA | AB189472.1 | - | 12 - 13 | 1434 | 58 |
AF168132.1 | - | 13 - 14 | 1747 | 58 | |
AK313965.1 | - | 13 - 14 | 1805 | 58 | |
AL080140.1 | - | 10 - 11 | 1088 | 58 | |
BC014614.2 | - | 13 - 14 | 1694 | 58 |