Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016329.3 | NP_057413.2 | 14 - 15 | 1861 | 103 |
XM_005265221.3 | XP_005265278.1 | 14 - 15 | 1977 | 103 | |
XM_006713203.3 | XP_006713266.1 | 15 - 16 | 2099 | 103 | |
XM_006713204.3 | XP_006713267.1 | 14 - 15 | 1799 | 103 | |
XM_011533824.2 | XP_011532126.1 | 15 - 16 | 1975 | 103 | |
XM_011533825.2 | XP_011532127.1 | 15 - 16 | 1822 | 103 | |
GenBank mRNA | AB189472.1 | - | 13 - 14 | 1485 | 103 |
AF168132.1 | - | 14 - 15 | 1798 | 103 | |
AK313965.1 | - | 14 - 15 | 1856 | 103 | |
AL080140.1 | - | 11 - 12 | 1139 | 103 | |
BC014614.2 | - | 14 - 15 | 1745 | 103 |