Assay Design



Gene Transcripts

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location
Chr.8: 61500556 - 61714640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164750.1 NP_001158222.1 12 - 13 965 159
NM_001164753.1 NP_001158225.1 12 - 13 881 159
NM_001164754.1 NP_001158226.1 11 - 12 1103 159
NM_001164755.1 NP_001158227.1 11 - 12 1031 159
NM_004318.3 NP_004309.2 12 - 13 1160 159
NM_032466.3 NP_115855.1 12 - 13 1160 159
NM_032468.4 NP_115857.1 13 - 14 1010 159
XM_005251235.2 XP_005251292.1 13 - 14 1174 159
XM_005251239.2 XP_005251296.1 12 - 13 1117 159
XM_005251240.1 XP_005251297.1 13 - 14 952 159
XM_005251242.2 XP_005251299.1 11 - 12 1072 159
XM_005251243.2 XP_005251300.1 12 - 13 1045 159
XM_005251246.2 XP_005251303.1 11 - 12 1000 159
XM_005251247.2 XP_005251304.1 11 - 12 988 159
XM_005251248.1 XP_005251305.1 11 - 12 758 159
XM_005251250.2 XP_005251307.1 10 - 11 943 159
XM_017013419.1 XP_016868908.1 13 - 14 1174 159
XM_017013422.1 XP_016868911.1 12 - 13 1129 159
XM_017013425.1 XP_016868914.1 12 - 13 1117 159
XM_017013426.1 XP_016868915.1 13 - 14 952 159
XM_017013428.1 XP_016868917.1 11 - 12 1072 159
XM_017013430.1 XP_016868919.1 12 - 13 1045 159
XM_017013431.1 XP_016868920.1 12 - 13 815 159
XM_017013437.1 XP_016868926.1 11 - 12 1000 159
XM_017013440.1 XP_016868929.1 12 - 13 823 159
XM_017013444.1 XP_016868933.1 11 - 12 686 159
XM_017013445.1 XP_016868934.1 12 - 13 815 159
XM_017013446.1 XP_016868935.1 11 - 12 758 159
XM_017013447.1 XP_016868936.1 11 - 12 686 159
GenBank mRNA AF289489.1 - 12 - 13 902 159
AF306765.1 - 13 - 14 931 159
AK291836.1 - 12 - 13 973 159
AK295528.1 - 12 - 13 845 159
AK298577.1 - 12 - 13 1024 159
AK304314.1 - 12 - 13 921 159
BC015518.2 - 11 - 12 1022 159
BC025236.1 - 11 - 12 959 159
EU718494.1 - 12 - 13 968 159
S83325.1 - 12 - 13 902 159
U03109.1 - 12 - 13 968 159

View Full Product Details