Assay Design



Gene Transcripts

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location
Chr.8: 61500556 - 61714640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164750.1 NP_001158222.1 22 - 23 1976 98
NM_004318.3 NP_004309.2 22 - 23 2171 98
XM_005251235.2 XP_005251292.1 23 - 24 2185 98
XM_005251236.2 XP_005251293.1 23 - 24 2182 98
XM_005251238.2 XP_005251295.1 22 - 23 2140 98
XM_005251239.2 XP_005251296.1 22 - 23 2128 98
XM_005251240.1 XP_005251297.1 23 - 24 1963 98
XM_005251242.2 XP_005251299.1 21 - 22 2083 98
XM_005251243.2 XP_005251300.1 22 - 23 2056 98
XM_005251244.1 XP_005251301.1 22 - 23 1903 98
XM_005251246.2 XP_005251303.1 21 - 22 2011 98
XM_005251247.2 XP_005251304.1 21 - 22 1999 98
XM_005251248.1 XP_005251305.1 21 - 22 1769 98
XM_005251250.2 XP_005251307.1 20 - 21 1954 98
XM_017013419.1 XP_016868908.1 23 - 24 2185 98
XM_017013420.1 XP_016868909.1 23 - 24 2182 98
XM_017013421.1 XP_016868910.1 22 - 23 2143 98
XM_017013422.1 XP_016868911.1 22 - 23 2140 98
XM_017013423.1 XP_016868912.1 22 - 23 2140 98
XM_017013424.1 XP_016868913.1 22 - 23 2137 98
XM_017013425.1 XP_016868914.1 22 - 23 2128 98
XM_017013426.1 XP_016868915.1 23 - 24 1963 98
XM_017013427.1 XP_016868916.1 23 - 24 1960 98
XM_017013428.1 XP_016868917.1 21 - 22 2083 98
XM_017013429.1 XP_016868918.1 22 - 23 2137 98
XM_017013430.1 XP_016868919.1 22 - 23 2056 98
XM_017013431.1 XP_016868920.1 22 - 23 1826 98
XM_017013432.1 XP_016868921.1 22 - 23 2053 98
XM_017013433.1 XP_016868922.1 22 - 23 1823 98
XM_017013434.1 XP_016868923.1 22 - 23 2125 98
XM_017013435.1 XP_016868924.1 22 - 23 1903 98
XM_017013436.1 XP_016868925.1 21 - 22 2098 98
XM_017013437.1 XP_016868926.1 21 - 22 2011 98
XM_017013438.1 XP_016868927.1 22 - 23 2053 98
XM_017013439.1 XP_016868928.1 22 - 23 1823 98
XM_017013440.1 XP_016868929.1 22 - 23 1834 98
XM_017013441.1 XP_016868930.1 21 - 22 2008 98
XM_017013442.1 XP_016868931.1 21 - 22 1996 98
XM_017013443.1 XP_016868932.1 20 - 21 1951 98
XM_017013444.1 XP_016868933.1 21 - 22 1697 98
GenBank mRNA AK304314.1 - 22 - 23 1932 98
S83325.1 - 22 - 23 1913 98
U03109.1 - 22 - 23 1976 98

View Full Product Details