Assay Design



Gene Transcripts

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location
Chr.8: 61500556 - 61714640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164750.1 NP_001158222.1 7 - 8 724 106
NM_001164751.1 NP_001158223.1 8 - 9 769 106
NM_004318.3 NP_004309.2 7 - 8 919 106
NM_032466.3 NP_115855.1 7 - 8 919 106
NM_032468.4 NP_115857.1 8 - 9 769 106
XM_005251235.2 XP_005251292.1 8 - 9 933 106
XM_005251236.2 XP_005251293.1 8 - 9 933 106
XM_005251238.2 XP_005251295.1 8 - 9 933 106
XM_005251240.1 XP_005251297.1 8 - 9 711 106
XM_017013419.1 XP_016868908.1 8 - 9 933 106
XM_017013420.1 XP_016868909.1 8 - 9 933 106
XM_017013421.1 XP_016868910.1 8 - 9 933 106
XM_017013422.1 XP_016868911.1 7 - 8 888 106
XM_017013423.1 XP_016868912.1 8 - 9 933 106
XM_017013424.1 XP_016868913.1 7 - 8 888 106
XM_017013426.1 XP_016868915.1 8 - 9 711 106
XM_017013427.1 XP_016868916.1 8 - 9 711 106
XM_017013429.1 XP_016868918.1 7 - 8 888 106
XM_017013431.1 XP_016868920.1 7 - 8 574 106
XM_017013433.1 XP_016868922.1 7 - 8 574 106
XM_017013436.1 XP_016868925.1 8 - 9 933 106
XM_017013439.1 XP_016868928.1 7 - 8 574 106
XM_017013445.1 XP_016868934.1 7 - 8 574 106
GenBank mRNA AF289489.1 - 7 - 8 661 106
AF306765.1 - 8 - 9 690 106
AK291836.1 - 7 - 8 732 106
AK304314.1 - 7 - 8 680 106
BC144362.1 - 8 - 9 699 106
BM785890.1 - 2 - 3 154 106
EU718494.1 - 7 - 8 727 106
S83325.1 - 7 - 8 661 106
U03109.1 - 7 - 8 727 106

View Full Product Details