Assay Design



Gene Transcripts

Gene Symbol
SLC5A7
Entrez Gene ID
60482
Gene Name
solute carrier family 5 member 7
Gene Aliases
CHT, CHT1, HMN7A
Chromosome Location
Chr.2: 107986514 - 108014497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305005.2 NP_001291934.1 3 - 4 589 101
NM_001305006.2 NP_001291935.1 2 - 3 363 101
NM_001305007.2 NP_001291936.1 3 - 4 592 101
NM_021815.4 NP_068587.1 3 - 4 592 101
GenBank mRNA AB043997.1 - 3 - 4 567 101
AF276871.1 - 2 - 3 291 101
AJ401466.1 - 2 - 3 309 101
AK300801.1 - 2 - 3 353 101
AK315280.1 - 3 - 4 581 101
BC111524.1 - 3 - 4 534 101
BC111525.1 - 3 - 4 539 101

View Full Product Details