Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000459.4 | NP_000450.2 | 8 - 9 | 1636 | 112 |
NM_001290077.1 | NP_001277006.1 | 7 - 8 | 1507 | 112 | |
NM_001290078.1 | NP_001277007.1 | 6 - 7 | 1195 | 112 | |
XM_005251561.1 | XP_005251618.1 | 8 - 9 | 1303 | 112 | |
XM_005251563.1 | XP_005251620.1 | 7 - 8 | 1174 | 112 | |
GenBank mRNA | AB086825.1 | - | 7 - 8 | 1163 | 112 |
AB208796.1 | - | 8 - 9 | 1314 | 112 | |
AK291775.1 | - | 8 - 9 | 1324 | 112 | |
AK294887.1 | - | 6 - 7 | 1196 | 112 | |
AK295043.1 | - | 7 - 8 | 1233 | 112 | |
AK308447.1 | - | 5 - 6 | 747 | 112 | |
BC035514.1 | - | 8 - 9 | 1311 | 112 | |
EU826592.1 | - | 7 - 8 | 1057 | 112 | |
L06139.1 | - | 8 - 9 | 1334 | 112 |