Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007215.3 | NP_009146.2 | 6 - 7 | 1293 | 80 |
GenBank mRNA | AF142992.1 | - | 6 - 7 | 1276 | 80 |
AF177201.1 | - | 6 - 7 | 1195 | 80 | |
AF184344.1 | - | 6 - 7 | 1276 | 80 | |
BC000913.2 | - | 6 - 7 | 1041 | 80 | |
BC009194.2 | - | 6 - 7 | 1263 | 80 | |
U94703.1 | - | 6 - 7 | 1277 | 80 |