Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162499.1 | NP_001155971.1 | 6 - 7 | 906 | 116 |
NM_012298.2 | NP_036430.1 | 4 - 5 | 627 | 116 | |
XM_011533503.2 | XP_011531805.1 | 6 - 7 | 924 | 116 | |
XM_011533504.2 | XP_011531806.1 | 6 - 7 | 1020 | 116 | |
GenBank mRNA | AB014567.1 | - | 4 - 5 | 555 | 116 |
BC136592.1 | - | 6 - 7 | 845 | 116 | |
DB471722.1 | - | 2 - 3 | 344 | 116 | |
GU727621.1 | - | 6 - 7 | 906 | 116 |