Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324066.1 | NP_001310995.1 | 3 - 4 | 719 | 134 |
NM_016641.3 | NP_057725.1 | 3 - 4 | 706 | 134 | |
NR_136689.1 | 3 - 4 | 723 | 134 | ||
GenBank mRNA | AF212862.1 | - | 3 - 4 | 662 | 134 |
AF306698.1 | - | 3 - 4 | 615 | 134 | |
AK312912.1 | - | 3 - 4 | 676 | 134 | |
AY463154.1 | - | 3 - 4 | 542 | 134 | |
BC014981.1 | - | 3 - 4 | 597 | 134 | |
BC025273.1 | - | 3 - 4 | 657 | 134 |