Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195098.1 | NP_001182027.1 | 10 - 11 | 1282 | 97 |
NM_001195099.1 | NP_001182028.1 | 8 - 9 | 973 | 97 | |
NM_015199.3 | NP_056014.2 | 10 - 11 | 1238 | 97 | |
XM_005264996.3 | XP_005265053.1 | 10 - 11 | 2609 | 97 | |
XM_011533540.2 | XP_011531842.1 | 10 - 11 | 2618 | 97 | |
XM_011533541.2 | XP_011531843.1 | 10 - 11 | 2618 | 97 | |
XM_011533542.2 | XP_011531844.1 | 9 - 10 | 1422 | 97 | |
XM_011533543.2 | XP_011531845.1 | 9 - 10 | 2417 | 97 | |
XM_011533545.2 | XP_011531847.1 | 10 - 11 | 1945 | 97 | |
XM_011533546.2 | XP_011531848.1 | 10 - 11 | 1936 | 97 | |
XM_011533547.2 | XP_011531849.1 | 9 - 10 | 1894 | 97 | |
XM_017006026.1 | XP_016861515.1 | 11 - 12 | 2016 | 97 | |
XM_017006027.1 | XP_016861516.1 | 10 - 11 | 1965 | 97 | |
XM_017006028.1 | XP_016861517.1 | 10 - 11 | 2618 | 97 | |
XM_017006029.1 | XP_016861518.1 | 9 - 10 | 1023 | 97 | |
GenBank mRNA | AB002377.2 | - | 10 - 11 | 1114 | 97 |
AK126888.1 | - | 10 - 11 | 1500 | 97 | |
AK293770.1 | - | 10 - 11 | 1283 | 97 | |
AK295813.1 | - | 9 - 10 | 1108 | 97 | |
AY367056.1 | - | 8 - 9 | 973 | 97 | |
BC106948.2 | - | 10 - 11 | 1312 | 97 | |
BC113868.1 | - | 10 - 11 | 1301 | 97 | |
BC114476.1 | - | 10 - 11 | 1312 | 97 |