Assay Design



Gene Transcripts

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
Chromosome Location
Chr.3: 15667236 - 15859814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195098.1 NP_001182027.1 12 - 13 1433 72
NM_001195099.1 NP_001182028.1 10 - 11 1124 72
NM_015199.3 NP_056014.2 12 - 13 1389 72
XM_005264996.3 XP_005265053.1 12 - 13 2760 72
XM_011533540.2 XP_011531842.1 12 - 13 2769 72
XM_011533541.2 XP_011531843.1 12 - 13 2769 72
XM_011533542.2 XP_011531844.1 11 - 12 1573 72
XM_011533543.2 XP_011531845.1 11 - 12 2568 72
XM_011533545.2 XP_011531847.1 12 - 13 2096 72
XM_011533546.2 XP_011531848.1 12 - 13 2087 72
XM_011533547.2 XP_011531849.1 11 - 12 2045 72
XM_017006026.1 XP_016861515.1 13 - 14 2167 72
XM_017006027.1 XP_016861516.1 12 - 13 2116 72
XM_017006028.1 XP_016861517.1 12 - 13 2769 72
XM_017006029.1 XP_016861518.1 11 - 12 1174 72
GenBank mRNA AB002377.2 - 12 - 13 1265 72
AK126888.1 - 12 - 13 1651 72
AK293770.1 - 12 - 13 1434 72
AK295813.1 - 11 - 12 1259 72
BC106948.2 - 12 - 13 1463 72
BC113868.1 - 12 - 13 1452 72
BC114476.1 - 12 - 13 1463 72

View Full Product Details