Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195098.1 | NP_001182027.1 | 13 - 14 | 1504 | 96 |
NM_001195099.1 | NP_001182028.1 | 11 - 12 | 1195 | 96 | |
NM_015199.3 | NP_056014.2 | 13 - 14 | 1460 | 96 | |
XM_005264996.3 | XP_005265053.1 | 13 - 14 | 2831 | 96 | |
XM_011533540.2 | XP_011531842.1 | 13 - 14 | 2840 | 96 | |
XM_011533541.2 | XP_011531843.1 | 13 - 14 | 2840 | 96 | |
XM_011533542.2 | XP_011531844.1 | 12 - 13 | 1644 | 96 | |
XM_011533543.2 | XP_011531845.1 | 12 - 13 | 2639 | 96 | |
XM_011533545.2 | XP_011531847.1 | 13 - 14 | 2167 | 96 | |
XM_011533546.2 | XP_011531848.1 | 13 - 14 | 2158 | 96 | |
XM_011533547.2 | XP_011531849.1 | 12 - 13 | 2116 | 96 | |
XM_017006026.1 | XP_016861515.1 | 14 - 15 | 2238 | 96 | |
XM_017006027.1 | XP_016861516.1 | 13 - 14 | 2187 | 96 | |
XM_017006028.1 | XP_016861517.1 | 13 - 14 | 2840 | 96 | |
XM_017006029.1 | XP_016861518.1 | 12 - 13 | 1245 | 96 | |
GenBank mRNA | AB002377.2 | - | 13 - 14 | 1336 | 96 |
AK126888.1 | - | 13 - 14 | 1722 | 96 | |
AK293770.1 | - | 13 - 14 | 1505 | 96 | |
AK295813.1 | - | 12 - 13 | 1330 | 96 | |
AY367056.1 | - | 11 - 12 | 1194 | 96 | |
BC106948.2 | - | 13 - 14 | 1534 | 96 | |
BC113868.1 | - | 13 - 14 | 1523 | 96 | |
BC114476.1 | - | 13 - 14 | 1534 | 96 |