Assay Design



Gene Transcripts

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
Chromosome Location
Chr.3: 15667236 - 15859814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195098.1 NP_001182027.1 19 - 20 2146 82
NM_001195099.1 NP_001182028.1 17 - 18 1837 82
NM_015199.3 NP_056014.2 19 - 20 2102 82
XM_005264996.3 XP_005265053.1 19 - 20 3473 82
XM_011533540.2 XP_011531842.1 19 - 20 3482 82
XM_011533541.2 XP_011531843.1 19 - 20 3482 82
XM_011533542.2 XP_011531844.1 18 - 19 2286 82
XM_011533543.2 XP_011531845.1 18 - 19 3281 82
XM_011533545.2 XP_011531847.1 19 - 20 2809 82
XM_011533546.2 XP_011531848.1 19 - 20 2800 82
XM_011533547.2 XP_011531849.1 18 - 19 2758 82
XM_017006026.1 XP_016861515.1 20 - 21 2880 82
XM_017006027.1 XP_016861516.1 19 - 20 2829 82
XM_017006029.1 XP_016861518.1 18 - 19 1887 82
GenBank mRNA AB002377.2 - 19 - 20 1978 82
AK293770.1 - 19 - 20 2147 82
AK295813.1 - 18 - 19 1972 82
AY367056.1 - 17 - 18 1836 82
BC106948.2 - 19 - 20 2176 82
BC113868.1 - 19 - 20 2165 82
BC114476.1 - 19 - 20 2176 82
BQ423818.1 - 5 - 6 518 82

View Full Product Details