Assay Design



Gene Transcripts

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
Chromosome Location
Chr.3: 15667236 - 15859814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195098.1 NP_001182027.1 21 - 22 2490 137
NM_001195099.1 NP_001182028.1 19 - 20 2181 137
NM_015199.3 NP_056014.2 21 - 22 2446 137
XM_005264996.3 XP_005265053.1 21 - 22 3817 137
XM_011533540.2 XP_011531842.1 21 - 22 3826 137
XM_011533541.2 XP_011531843.1 21 - 22 3826 137
XM_011533542.2 XP_011531844.1 20 - 21 2630 137
XM_011533543.2 XP_011531845.1 20 - 21 3625 137
XM_011533545.2 XP_011531847.1 21 - 22 3153 137
XM_011533546.2 XP_011531848.1 21 - 22 3144 137
XM_011533547.2 XP_011531849.1 20 - 21 3102 137
XM_017006026.1 XP_016861515.1 22 - 23 3224 137
XM_017006027.1 XP_016861516.1 21 - 22 3173 137
XM_017006029.1 XP_016861518.1 20 - 21 2231 137
GenBank mRNA AB002377.2 - 21 - 22 2322 137
AK293770.1 - 21 - 22 2491 137
AY367056.1 - 19 - 20 2180 137

View Full Product Details