Assay Design



Gene Transcripts

Gene Symbol
ANKRD28
Entrez Gene ID
23243
Gene Name
ankyrin repeat domain 28
Gene Aliases
PITK, PPP1R65
Chromosome Location
Chr.3: 15667236 - 15859814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195098.1 NP_001182027.1 26 - 27 2975 125
NM_001195099.1 NP_001182028.1 24 - 25 2666 125
NM_015199.3 NP_056014.2 26 - 27 2931 125
XM_005264996.3 XP_005265053.1 26 - 27 4302 125
XM_011533540.2 XP_011531842.1 26 - 27 4311 125
XM_011533541.2 XP_011531843.1 26 - 27 4311 125
XM_011533542.2 XP_011531844.1 25 - 26 3115 125
XM_011533543.2 XP_011531845.1 25 - 26 4110 125
XM_011533545.2 XP_011531847.1 26 - 27 3638 125
XM_011533546.2 XP_011531848.1 26 - 27 3629 125
XM_011533547.2 XP_011531849.1 25 - 26 3587 125
XM_017006026.1 XP_016861515.1 27 - 28 3709 125
XM_017006027.1 XP_016861516.1 26 - 27 3658 125
XM_017006029.1 XP_016861518.1 25 - 26 2716 125
GenBank mRNA AB002377.2 - 26 - 27 2807 125
AK293770.1 - 26 - 27 2976 125
AY367056.1 - 24 - 25 2665 125

View Full Product Details