Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020397.3 | NP_065130.1 | 5 - 6 | 843 | 90 |
NM_153498.3 | NP_705718.1 | 5 - 6 | 843 | 90 | |
XM_006717481.3 | XP_006717544.1 | 5 - 6 | 1107 | 90 | |
XM_006717482.3 | XP_006717545.1 | 5 - 6 | 872 | 90 | |
XM_006717483.3 | XP_006717546.1 | 5 - 6 | 834 | 90 | |
XM_011519591.2 | XP_011517893.1 | 5 - 6 | 1280 | 90 | |
XM_011519592.2 | XP_011517894.1 | 4 - 5 | 1472 | 90 | |
XM_011519593.2 | XP_011517895.1 | 4 - 5 | 1520 | 90 | |
XM_011519594.2 | XP_011517896.1 | 3 - 4 | 461 | 90 | |
XM_011519595.2 | XP_011517897.1 | 4 - 5 | 1334 | 90 | |
XM_017016438.1 | XP_016871927.1 | 6 - 7 | 1138 | 90 | |
XM_017016439.1 | XP_016871928.1 | 7 - 8 | 969 | 90 | |
GenBank mRNA | AB081726.1 | - | 5 - 6 | 799 | 90 |
AF286366.1 | - | 5 - 6 | 654 | 90 | |
AK289393.1 | - | 6 - 7 | 654 | 90 | |
BC035745.1 | - | 5 - 6 | 660 | 90 |