Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000700.2 | NP_000691.1 | 6 - 7 | 680 | 121 |
XM_011518609.1 | XP_011516911.1 | 6 - 7 | 607 | 121 | |
XM_017014657.1 | XP_016870146.1 | 6 - 7 | 626 | 121 | |
GenBank mRNA | AB451274.1 | - | 5 - 6 | 472 | 121 |
AB451401.1 | - | 5 - 6 | 472 | 121 | |
AK074480.1 | - | 5 - 6 | 1354 | 121 | |
AK296808.1 | - | 6 - 7 | 595 | 121 | |
BC001275.1 | - | 6 - 7 | 524 | 121 | |
BC035993.1 | - | 6 - 7 | 546 | 121 | |
BT019896.1 | - | 5 - 6 | 472 | 121 | |
BT019916.1 | - | 5 - 6 | 472 | 121 | |
BT019917.1 | - | 5 - 6 | 472 | 121 | |
CR407684.1 | - | 5 - 6 | 472 | 121 | |
X05908.1 | - | 6 - 7 | 546 | 121 |