Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128620.1 | NP_001122092.1 | 5 - 6 | 1013 | 100 |
NM_002576.4 | NP_002567.3 | 5 - 6 | 1013 | 100 | |
XM_011545080.2 | XP_011543382.1 | 5 - 6 | 720 | 100 | |
XM_011545081.2 | XP_011543383.1 | 5 - 6 | 789 | 100 | |
XM_011545082.2 | XP_011543384.1 | 6 - 7 | 743 | 100 | |
XM_011545083.2 | XP_011543385.1 | 5 - 6 | 1010 | 100 | |
XM_011545084.2 | XP_011543386.1 | 5 - 6 | 580 | 100 | |
XM_017017846.1 | XP_016873335.1 | 5 - 6 | 522 | 100 | |
XM_017017847.1 | XP_016873336.1 | 5 - 6 | 720 | 100 | |
XM_017017848.1 | XP_016873337.1 | 5 - 6 | 789 | 100 | |
XM_017017849.1 | XP_016873338.1 | 5 - 6 | 666 | 100 | |
XM_017017850.1 | XP_016873339.1 | 6 - 7 | 729 | 100 | |
GenBank mRNA | AF071884.1 | - | 4 - 5 | 491 | 100 |
AK055228.1 | - | 5 - 6 | 680 | 100 | |
AK293098.1 | - | 5 - 6 | 669 | 100 | |
AK296006.1 | - | 4 - 5 | 421 | 100 | |
BC050377.1 | - | 5 - 6 | 605 | 100 | |
BC109298.1 | - | 5 - 6 | 621 | 100 | |
BC109299.1 | - | 5 - 6 | 621 | 100 | |
U24152.1 | - | 5 - 6 | 873 | 100 | |
U51120.1 | - | 5 - 6 | 534 | 100 |