Assay Design



Gene Transcripts

Gene Symbol
CPXCR1
Entrez Gene ID
53336
Gene Name
CPX chromosome region, candidate 1
Gene Aliases
CT77
Chromosome Location
Chr.X: 88747225 - 88754785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184771.1 NP_001171700.1 3 - 3 1029 151
NM_033048.5 NP_149037.4 3 - 3 1067 151
GenBank mRNA AF284767.1 - 1 - 1 844 151
AK098646.1 - 3 - 3 1004 151
BC027614.2 - 3 - 3 1037 151

View Full Product Details