Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130823.2 | NP_001124295.1 | 25 - 26 | 2619 | 100 |
NM_001318730.1 | NP_001305659.1 | 24 - 25 | 2490 | 100 | |
NM_001318731.1 | NP_001305660.1 | 25 - 26 | 2498 | 100 | |
NM_001379.3 | NP_001370.1 | 24 - 25 | 2571 | 100 | |
GenBank mRNA | AB209413.2 | - | 24 - 25 | 2297 | 100 |
AF180682.1 | - | 22 - 23 | 2078 | 100 | |
BC092517.1 | - | 25 - 26 | 2378 | 100 | |
BC126227.1 | - | 25 - 26 | 2563 | 100 | |
BC144093.1 | - | 25 - 26 | 2563 | 100 | |
X63692.1 | - | 24 - 25 | 2572 | 100 |