Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130823.2 | NP_001124295.1 | 33 - 34 | 4044 | 118 |
NM_001318730.1 | NP_001305659.1 | 32 - 33 | 3915 | 118 | |
NM_001318731.1 | NP_001305660.1 | 33 - 34 | 3923 | 118 | |
NM_001379.3 | NP_001370.1 | 32 - 33 | 3996 | 118 | |
GenBank mRNA | AB209413.2 | - | 32 - 33 | 3722 | 118 |
AF180682.1 | - | 30 - 31 | 3503 | 118 | |
AK122759.1 | - | 5 - 6 | 1680 | 118 | |
BC092517.1 | - | 33 - 34 | 3803 | 118 | |
BC126227.1 | - | 33 - 34 | 3988 | 118 | |
BC144093.1 | - | 33 - 34 | 3988 | 118 | |
X63692.1 | - | 32 - 33 | 3997 | 118 |