Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003204.2 | NP_003195.1 | 3 - 4 | 1371 | 97 |
XM_005257410.3 | XP_005257467.1 | 3 - 4 | 1317 | 97 | |
XM_005257411.3 | XP_005257468.1 | 3 - 4 | 1284 | 97 | |
XM_017024691.1 | XP_016880180.1 | 3 - 4 | 1338 | 97 | |
GenBank mRNA | AK090459.1 | - | 3 - 4 | 1336 | 97 |
BC131769.1 | - | 2 - 3 | 964 | 97 | |
X77366.1 | - | 3 - 4 | 1312 | 97 |