Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000770.3 | NP_000761.3 | 2 - 3 | 425 | 80 |
NM_001198853.1 | NP_001185782.1 | 2 - 3 | 463 | 80 | |
NM_001198855.1 | NP_001185784.1 | 3 - 4 | 525 | 80 | |
GenBank mRNA | AK292753.1 | - | 2 - 3 | 425 | 80 |
AK293327.1 | - | 2 - 3 | 463 | 80 | |
AK315823.1 | - | 2 - 3 | 425 | 80 | |
BC020596.1 | - | 2 - 3 | 425 | 80 | |
CD014119.1 | - | 3 - 4 | 514 | 80 | |
M17397.1 | - | 2 - 3 | 331 | 80 | |
M17398.1 | - | 2 - 3 | 331 | 80 | |
M21941.1 | - | 2 - 3 | 300 | 80 | |
Y00498.1 | - | 2 - 3 | 372 | 80 |