Assay Design



Gene Transcripts

Gene Symbol
SMARCA4
Entrez Gene ID
6597
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
Gene Aliases
BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b
Chromosome Location
Chr.19: 10960922 - 11062282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128844.1 NP_001122316.1 13 - 14 2297 96
NM_001128845.1 NP_001122317.1 11 - 12 1937 96
NM_001128846.1 NP_001122318.1 11 - 12 1937 96
NM_001128847.1 NP_001122319.1 11 - 12 1937 96
NM_001128848.1 NP_001122320.1 11 - 12 1937 96
NM_001128849.1 NP_001122321.1 12 - 13 2011 96
NM_003072.3 NP_003063.2 12 - 13 2221 96
XM_006722845.2 XP_006722908.1 12 - 13 2143 96
XM_006722846.2 XP_006722909.1 12 - 13 2237 96
XM_011528198.1 XP_011526500.1 13 - 14 2218 96
XM_017027160.1 XP_016882649.1 13 - 14 2218 96
XM_017027161.1 XP_016882650.1 13 - 14 2218 96
XM_017027162.1 XP_016882651.1 13 - 14 2218 96
XM_017027163.1 XP_016882652.1 13 - 14 2218 96
XM_017027164.1 XP_016882653.1 13 - 14 2218 96
XM_017027165.1 XP_016882654.1 13 - 14 2218 96
XM_017027166.1 XP_016882655.1 13 - 14 2218 96
XM_017027167.1 XP_016882656.1 13 - 14 2218 96
XM_017027168.1 XP_016882657.1 13 - 14 2218 96
GenBank mRNA AB209313.2 - 11 - 12 1811 96
AK290033.1 - 13 - 14 2218 96
BC136644.1 - 10 - 11 2157 96
BC150298.1 - 12 - 13 2142 96
D26156.1 - 12 - 13 1994 96
EU430756.1 - 11 - 12 1937 96
EU430757.1 - 11 - 12 1937 96
EU430758.1 - 11 - 12 1937 96
EU430759.1 - 11 - 12 1937 96
U29175.1 - 12 - 13 2015 96

View Full Product Details