Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128844.1 | NP_001122316.1 | 15 - 16 | 2484 | 58 |
NM_001128845.1 | NP_001122317.1 | 13 - 14 | 2124 | 58 | |
NM_001128846.1 | NP_001122318.1 | 13 - 14 | 2124 | 58 | |
NM_001128847.1 | NP_001122319.1 | 13 - 14 | 2124 | 58 | |
NM_001128848.1 | NP_001122320.1 | 13 - 14 | 2124 | 58 | |
NM_001128849.1 | NP_001122321.1 | 14 - 15 | 2198 | 58 | |
NM_003072.3 | NP_003063.2 | 14 - 15 | 2408 | 58 | |
XM_006722845.2 | XP_006722908.1 | 14 - 15 | 2330 | 58 | |
XM_006722846.2 | XP_006722909.1 | 14 - 15 | 2424 | 58 | |
XM_011528198.1 | XP_011526500.1 | 15 - 16 | 2405 | 58 | |
XM_017027160.1 | XP_016882649.1 | 15 - 16 | 2405 | 58 | |
XM_017027161.1 | XP_016882650.1 | 15 - 16 | 2405 | 58 | |
XM_017027162.1 | XP_016882651.1 | 15 - 16 | 2405 | 58 | |
XM_017027163.1 | XP_016882652.1 | 15 - 16 | 2405 | 58 | |
XM_017027164.1 | XP_016882653.1 | 15 - 16 | 2405 | 58 | |
XM_017027165.1 | XP_016882654.1 | 15 - 16 | 2405 | 58 | |
XM_017027166.1 | XP_016882655.1 | 15 - 16 | 2405 | 58 | |
XM_017027167.1 | XP_016882656.1 | 15 - 16 | 2405 | 58 | |
XM_017027168.1 | XP_016882657.1 | 15 - 16 | 2405 | 58 | |
GenBank mRNA | AB209313.2 | - | 13 - 14 | 1998 | 58 |
AK290033.1 | - | 15 - 16 | 2405 | 58 | |
BC136644.1 | - | 12 - 13 | 2344 | 58 | |
BC150298.1 | - | 14 - 15 | 2329 | 58 | |
D26156.1 | - | 14 - 15 | 2181 | 58 | |
EU430756.1 | - | 13 - 14 | 2124 | 58 | |
EU430757.1 | - | 13 - 14 | 2124 | 58 | |
EU430758.1 | - | 13 - 14 | 2124 | 58 | |
EU430759.1 | - | 13 - 14 | 2124 | 58 | |
U29175.1 | - | 14 - 15 | 2202 | 58 |