Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128844.1 | NP_001122316.1 | 18 - 19 | 2870 | 57 |
NM_001128845.1 | NP_001122317.1 | 16 - 17 | 2510 | 57 | |
NM_001128846.1 | NP_001122318.1 | 16 - 17 | 2510 | 57 | |
NM_001128847.1 | NP_001122319.1 | 16 - 17 | 2510 | 57 | |
NM_001128848.1 | NP_001122320.1 | 16 - 17 | 2510 | 57 | |
NM_001128849.1 | NP_001122321.1 | 17 - 18 | 2584 | 57 | |
NM_003072.3 | NP_003063.2 | 17 - 18 | 2794 | 57 | |
XM_006722845.2 | XP_006722908.1 | 17 - 18 | 2716 | 57 | |
XM_006722846.2 | XP_006722909.1 | 17 - 18 | 2810 | 57 | |
XM_011528198.1 | XP_011526500.1 | 18 - 19 | 2791 | 57 | |
XM_017027160.1 | XP_016882649.1 | 18 - 19 | 2791 | 57 | |
XM_017027161.1 | XP_016882650.1 | 18 - 19 | 2791 | 57 | |
XM_017027162.1 | XP_016882651.1 | 18 - 19 | 2791 | 57 | |
XM_017027163.1 | XP_016882652.1 | 18 - 19 | 2791 | 57 | |
XM_017027164.1 | XP_016882653.1 | 18 - 19 | 2791 | 57 | |
XM_017027165.1 | XP_016882654.1 | 18 - 19 | 2791 | 57 | |
XM_017027166.1 | XP_016882655.1 | 18 - 19 | 2791 | 57 | |
XM_017027167.1 | XP_016882656.1 | 18 - 19 | 2791 | 57 | |
XM_017027168.1 | XP_016882657.1 | 18 - 19 | 2791 | 57 | |
GenBank mRNA | AB209313.2 | - | 16 - 17 | 2384 | 57 |
AK290033.1 | - | 18 - 19 | 2791 | 57 | |
AK299761.1 | - | 3 - 4 | 252 | 57 | |
AK303352.1 | - | 2 - 3 | 194 | 57 | |
AK308763.1 | - | 2 - 3 | 423 | 57 | |
BC136644.1 | - | 15 - 16 | 2730 | 57 | |
BC150298.1 | - | 17 - 18 | 2715 | 57 | |
D26156.1 | - | 17 - 18 | 2567 | 57 | |
EU430756.1 | - | 16 - 17 | 2510 | 57 | |
EU430757.1 | - | 16 - 17 | 2510 | 57 | |
EU430758.1 | - | 16 - 17 | 2510 | 57 | |
EU430759.1 | - | 16 - 17 | 2510 | 57 | |
U29175.1 | - | 17 - 18 | 2588 | 57 |