Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128844.1 | NP_001122316.1 | 20 - 21 | 3220 | 67 |
NM_001128845.1 | NP_001122317.1 | 18 - 19 | 2860 | 67 | |
NM_001128846.1 | NP_001122318.1 | 18 - 19 | 2860 | 67 | |
NM_001128847.1 | NP_001122319.1 | 18 - 19 | 2860 | 67 | |
NM_001128848.1 | NP_001122320.1 | 18 - 19 | 2860 | 67 | |
NM_001128849.1 | NP_001122321.1 | 19 - 20 | 2934 | 67 | |
NM_003072.3 | NP_003063.2 | 19 - 20 | 3144 | 67 | |
XM_006722845.2 | XP_006722908.1 | 19 - 20 | 3066 | 67 | |
XM_006722846.2 | XP_006722909.1 | 19 - 20 | 3160 | 67 | |
XM_011528198.1 | XP_011526500.1 | 20 - 21 | 3141 | 67 | |
XM_017027160.1 | XP_016882649.1 | 20 - 21 | 3141 | 67 | |
XM_017027161.1 | XP_016882650.1 | 20 - 21 | 3141 | 67 | |
XM_017027162.1 | XP_016882651.1 | 20 - 21 | 3141 | 67 | |
XM_017027163.1 | XP_016882652.1 | 20 - 21 | 3141 | 67 | |
XM_017027164.1 | XP_016882653.1 | 20 - 21 | 3141 | 67 | |
XM_017027165.1 | XP_016882654.1 | 20 - 21 | 3141 | 67 | |
XM_017027166.1 | XP_016882655.1 | 20 - 21 | 3141 | 67 | |
XM_017027167.1 | XP_016882656.1 | 20 - 21 | 3141 | 67 | |
XM_017027168.1 | XP_016882657.1 | 20 - 21 | 3141 | 67 | |
GenBank mRNA | AB209313.2 | - | 18 - 19 | 2734 | 67 |
AK026573.1 | - | 1 - 2 | 239 | 67 | |
AK055168.1 | - | 2 - 3 | 932 | 67 | |
AK097105.1 | - | 1 - 2 | 1389 | 67 | |
AK290033.1 | - | 20 - 21 | 3141 | 67 | |
AK299761.1 | - | 5 - 6 | 602 | 67 | |
AK303352.1 | - | 4 - 5 | 544 | 67 | |
AK308763.1 | - | 4 - 5 | 773 | 67 | |
BC136644.1 | - | 17 - 18 | 3080 | 67 | |
BC150298.1 | - | 19 - 20 | 3065 | 67 | |
D26156.1 | - | 19 - 20 | 2917 | 67 | |
EU430756.1 | - | 18 - 19 | 2860 | 67 | |
EU430757.1 | - | 18 - 19 | 2860 | 67 | |
EU430758.1 | - | 18 - 19 | 2860 | 67 | |
EU430759.1 | - | 18 - 19 | 2860 | 67 | |
U29175.1 | - | 19 - 20 | 2938 | 67 |