Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128844.1 | NP_001122316.1 | 28 - 29 | 4239 | 84 |
NM_001128849.1 | NP_001122321.1 | 27 - 28 | 3953 | 84 | |
NM_003072.3 | NP_003063.2 | 27 - 28 | 4163 | 84 | |
XM_006722845.2 | XP_006722908.1 | 27 - 28 | 4085 | 84 | |
XM_006722846.2 | XP_006722909.1 | 27 - 28 | 4179 | 84 | |
XM_011528198.1 | XP_011526500.1 | 28 - 29 | 4160 | 84 | |
XM_017027160.1 | XP_016882649.1 | 28 - 29 | 4160 | 84 | |
XM_017027161.1 | XP_016882650.1 | 28 - 29 | 4160 | 84 | |
XM_017027163.1 | XP_016882652.1 | 28 - 29 | 4160 | 84 | |
GenBank mRNA | AK290033.1 | - | 28 - 29 | 4160 | 84 |
AK299761.1 | - | 13 - 14 | 1621 | 84 | |
BC150298.1 | - | 27 - 28 | 4084 | 84 | |
D26156.1 | - | 27 - 28 | 3936 | 84 | |
U29175.1 | - | 27 - 28 | 3957 | 84 |