Assay Design
Gene Transcripts
- Gene Symbol
- SMARCA4
- Entrez Gene ID
-
6597
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
- Gene Aliases
-
BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b
- Chromosome Location
-
Chr.19: 10960922 - 11062282 on Build GRCh38
View Full Product Details