Assay Design



Gene Transcripts

Gene Symbol
SMARCA4
Entrez Gene ID
6597
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
Gene Aliases
BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b
Chromosome Location
Chr.19: 10960922 - 11062282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128844.1 NP_001122316.1 9 - 10 1781 89
NM_001128845.1 NP_001122317.1 7 - 8 1421 89
NM_001128846.1 NP_001122318.1 7 - 8 1421 89
NM_001128847.1 NP_001122319.1 7 - 8 1421 89
NM_001128848.1 NP_001122320.1 7 - 8 1421 89
NM_001128849.1 NP_001122321.1 8 - 9 1495 89
NM_003072.3 NP_003063.2 8 - 9 1705 89
XM_006722845.2 XP_006722908.1 8 - 9 1627 89
XM_006722846.2 XP_006722909.1 8 - 9 1721 89
XM_011528198.1 XP_011526500.1 9 - 10 1702 89
XM_017027160.1 XP_016882649.1 9 - 10 1702 89
XM_017027161.1 XP_016882650.1 9 - 10 1702 89
XM_017027162.1 XP_016882651.1 9 - 10 1702 89
XM_017027163.1 XP_016882652.1 9 - 10 1702 89
XM_017027164.1 XP_016882653.1 9 - 10 1702 89
XM_017027165.1 XP_016882654.1 9 - 10 1702 89
XM_017027166.1 XP_016882655.1 9 - 10 1702 89
XM_017027167.1 XP_016882656.1 9 - 10 1702 89
XM_017027168.1 XP_016882657.1 9 - 10 1702 89
GenBank mRNA AB209313.2 - 7 - 8 1295 89
AK290033.1 - 9 - 10 1702 89
BC035916.1 - 8 - 9 1555 89
BC136644.1 - 7 - 8 1449 89
BC139917.1 - 8 - 9 1594 89
BC139924.1 - 8 - 9 1594 89
BC150298.1 - 8 - 9 1626 89
D26156.1 - 8 - 9 1478 89
EU430756.1 - 7 - 8 1421 89
EU430757.1 - 7 - 8 1421 89
EU430758.1 - 7 - 8 1421 89
EU430759.1 - 7 - 8 1421 89
U29175.1 - 8 - 9 1499 89

View Full Product Details