Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270934.1 | NP_001257863.1 | 17 - 18 | 2623 | 141 |
NM_001270941.1 | NP_001257870.1 | 18 - 19 | 2686 | 141 | |
NM_001282282.1 | NP_001269211.1 | 17 - 18 | 2409 | 141 | |
NM_014790.4 | NP_055605.2 | 18 - 19 | 2686 | 141 | |
XM_017010097.1 | XP_016865586.1 | 19 - 20 | 2680 | 141 | |
XM_017010098.1 | XP_016865587.1 | 17 - 18 | 2566 | 141 | |
GenBank mRNA | AB011127.1 | - | 17 - 18 | 2317 | 141 |
AF273057.1 | - | 18 - 19 | 2527 | 141 | |
AK291280.1 | - | 18 - 19 | 2408 | 141 | |
AK299724.1 | - | 17 - 18 | 2335 | 141 | |
BC017354.2 | - | 18 - 19 | 2451 | 141 | |
EF512550.1 | - | 17 - 18 | 2145 | 141 |